Skip to Content
Merck
All Photos(1)

Key Documents

744859

Sigma-Aldrich

(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium triflate

≥98.0% (HPLC)

Synonym(s):

(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate, Mesityl(p-tolyl)iodonium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18F3IO3S
CAS Number:
Molecular Weight:
486.29
Beilstein:
20040208
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

form

powder

composition

carbon content, 41.2-42.8%

functional group

fluoro
iodo
triflate

SMILES string

[O-]S(=O)(=O)C(F)(F)F.Cc1ccc([I+]c2c(C)cc(C)cc2C)cc1

InChI

1S/C16H18I.CHF3O3S/c1-11-5-7-15(8-6-11)17-16-13(3)9-12(2)10-14(16)4;2-1(3,4)8(5,6)7/h5-10H,1-4H3;(H,5,6,7)/q+1;/p-1

InChI key

KVLSSMIQFXWHII-UHFFFAOYSA-M

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service