Skip to Content
Merck
All Photos(1)

Key Documents

743569

Sigma-Aldrich

Bis[(10,11-η)-5-[(11bS)-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-yl- κP4]-5H-dibenz[b,f]azepine]rhodium(I) tetrafluoroborate salt

97%

Synonym(s):

Bis[(10,11-η)-(S)-N-[(3,5-dioxa-4-phosphacyclohepta[2,1-a;3,4-a′]dinaphthalen-4-yl-κP4)]-dibenzo[b,f]azepine]rhodium(I) tetrafluoroborate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C68H44BF4N2O4P2Rh
CAS Number:
Molecular Weight:
1204.75
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97% (elemental analysis)
97%

composition

carbon content, 68.42-70.16%

reaction suitability

core: rhodium
reagent type: catalyst

SMILES string

[Rh+].F[B-](F)(F)F.O1P(Oc2ccc3ccccc3c2-c4c1ccc5ccccc45)N6c7ccccc7C=Cc8ccccc68.O9P(Oc%10ccc%11ccccc%11c%10-c%12c9ccc%13ccccc%12%13)N%14c%15ccccc%15C=Cc%16ccccc%14%16

InChI

1S/2C34H22NO2P.BF4.Rh/c2*1-5-13-27-23(9-1)19-21-31-33(27)34-28-14-6-2-10-24(28)20-22-32(34)37-38(36-31)35-29-15-7-3-11-25(29)17-18-26-12-4-8-16-30(26)35;2-1(3,4)5;/h2*1-22H;;/q;;-1;+1

InChI key

RFMQVNMFBNKHLB-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

contains ~10% ether and 5% THF

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service