Skip to Content
Merck
All Photos(1)

Key Documents

739804

Sigma-Aldrich

6-(Trifluoromethyl)pyridine-2-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
£44.31

£44.31

List price£63.30Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
£44.31

About This Item

Empirical Formula (Hill Notation):
C7H4F3NO2
Molecular Weight:
191.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

£44.31

List price£63.30Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

154-158 °C

functional group

carboxylic acid
fluoro

storage temp.

2-8°C

SMILES string

OC(=O)c1cccc(n1)C(F)(F)F

InChI

1S/C7H4F3NO2/c8-7(9,10)5-3-1-2-4(11-5)6(12)13/h1-3H,(H,12,13)

InChI key

OKBHXGBLXDNJJD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service