Skip to Content
Merck
All Photos(1)

Documents

736848

Sigma-Aldrich

Chlorotris(triethylsilyl)silane

Synonym(s):

Supersilyl TES

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H45ClSi4
CAS Number:
Molecular Weight:
409.34
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.535

density

0.941 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC[Si](CC)(CC)[Si](Cl)([Si](CC)(CC)CC)[Si](CC)(CC)CC

InChI

1S/C18H45ClSi4/c1-10-20(11-2,12-3)23(19,21(13-4,14-5)15-6)22(16-7,17-8)18-9/h10-18H2,1-9H3

InChI key

PWDJKBJJIRTHAW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Yamamoto group has employed several applications in catalysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service