Skip to Content
Merck
All Photos(2)

Key Documents

730564

Sigma-Aldrich

2,3-Dichloro-4-iodopyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2Cl2IN
CAS Number:
Molecular Weight:
273.89
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

109-113 °C

functional group

chloro
iodo

SMILES string

Clc1nccc(I)c1Cl

InChI

1S/C5H2Cl2IN/c6-4-3(8)1-2-9-5(4)7/h1-2H

InChI key

WRPBPDPXVUBWCR-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Zhao et al.
The Journal of organic chemistry, 73(17), 6580-6586 (2008-08-08)
A new approach wherein steric interactions between substituents of unsymmetrical bis(4-pyridyl)acetylene ligands dictate the self-selection of single isomers of [4 + 4] self-assembled squares is presented. Each [4 + 4] self-assembly is characterized by multinuclear (31)P and (1)H NMR spectroscopies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service