Skip to Content
Merck
All Photos(1)

Key Documents

728705

Sigma-Aldrich

MK-2 Dye

95%

Synonym(s):

2-Cyano-3-[5′′′-(9-ethyl-9H-carbazol-3-yl)-3′,3′′,3′′′,4-tetra-n-hexyl-[2,2′,5′,2′′,5′′,2′′′]-quater thiophen-5-yl] acrylic acid, metal free solar cell dye

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H70N2O2S4
CAS Number:
Molecular Weight:
955.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

mp

181-185 °C

storage temp.

2-8°C

SMILES string

CCCCCCc1cc(sc1\C=C(/C#N)C(O)=O)-c2sc(cc2CCCCCC)-c3sc(cc3CCCCCC)-c4sc(cc4CCCCCC)-c5ccc6n(CC)c7ccccc7c6c5

InChI

1S/C58H70N2O2S4/c1-6-11-15-19-25-40-34-52(63-50(40)38-45(39-59)58(61)62)55-43(27-21-17-13-8-3)36-54(65-55)57-44(28-22-18-14-9-4)37-53(66-57)56-42(26-20-16-12-7-2)35-51(64-56)41-31-32-49-47(33-41)46-29-23-24-30-48(46)60(49)10-5/h23-24,29-38H,6-22,25-28H2,1-5H3,(H,61,62)/b45-38+

InChI key

FOELOZKDLHJOHT-XLDZHHEVSA-N

General description

MK-2 Dye is an oligothiophene based organic dye that can be used as a photosensitizer. It has a tetrameric oligothiophene spacer with donor and acceptor units. It also shows a conversion efficiency of 8%.

Application

MK-2 Dye is an alkyl functionalized dye which can be used in a variety of applications such as dye sensitized solar cells and photovoltaic cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Synthesis of a Regioregular Oligothiophene Photovoltaic Dye Molecule, MK-2, and Related Compounds: A Cooperative Hypervalent Iodine and Metal-Catalyzed Synthetic Route
Dohi T, et al.
Chemistry?A European Journal , 19(6), 2067-2075 (2013)
Alkyl-functionalized organic dyes for efficient molecular photovoltaics
Koumura N, et al.
Journal of the American Chemical Society, 128(44), 14256-14257 (2006)
Wang, Z. S.; Koumura, N.; Cui, Y.;
Chemistry of Materials, 20, 3993-3993 (2008)
Highly stable sensitizer dyes for dye-sensitized solar cells: role of the oligothiophene moiety
Katoh R, et al.
Energy & Environmental Science, 2(5), 542-546 (2009)
Efficient low-temperature transparent electrocatalytic layers based on graphene oxide nanosheets for dye-sensitized solar cells
Seo SH, et al.
ACS Applied Materials & Interfaces, 7(20), 10863-10871 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service