Skip to Content
Merck
All Photos(2)

Key Documents

720348

Sigma-Aldrich

2-Chloro-5-nitropyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2ClN3O2
CAS Number:
Molecular Weight:
159.53
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

105-110 °C

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1cnc(Cl)nc1

InChI

1S/C4H2ClN3O2/c5-4-6-1-3(2-7-4)8(9)10/h1-2H

InChI key

OFCBNMYNAHUDGE-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola R Campodónico et al.
Frontiers in chemistry, 8, 583-583 (2020-08-06)
The mechanism of SNAr reactions between 2-chloro-5-nitropyrimidine with primary and secondary alicyclic amines, respectively, have been studied by kinetic measurements. The kinetic data obtained in aqueous media opens a controversial discussion based on Brönsted-type plots analysis. The first approach based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service