Skip to Content
Merck
All Photos(1)

Key Documents

696943

Sigma-Aldrich

(1S,2S)-2-[(4R,11bS)-3H-dinaphtho[2,1-c:1′,2′-e]phosphepin-4(5H)-yl]-1,2-diphenylethylamine

kanata purity

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H30NP
Molecular Weight:
507.60
UNSPSC Code:
12352005
PubChem Substance ID:

form

solid

optical activity

[α]22/D +162°, c = 0.5 in chloroform

mp

91-96 °C

functional group

amine
phenyl
phosphine

SMILES string

N[C@H]([C@@H](P1Cc2ccc3ccccc3c2-c4c(C1)ccc5ccccc45)c6ccccc6)c7ccccc7

InChI

1S/C36H30NP/c37-35(27-13-3-1-4-14-27)36(28-15-5-2-6-16-28)38-23-29-21-19-25-11-7-9-17-31(25)33(29)34-30(24-38)22-20-26-12-8-10-18-32(26)34/h1-22,35-36H,23-24,37H2/t35-,36-/m0/s1

InChI key

YHBAPPMVVCHAQC-ZPGRZCPFSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Aminophosphine Ligands

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service