Skip to Content
Merck
All Photos(1)

Key Documents

696056

Sigma-Aldrich

Iodoethane-13C2,d5

99 atom % 13C, 98 atom % D, 98% (CP)

Synonym(s):

Ethyl-13C2,15N iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CD313CD2I
Molecular Weight:
162.98
UNSPSC Code:
12352108

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
98 atom % D

Assay

98% (CP)

form

liquid

bp

69-73 °C (lit.)

mp

-108 °C (lit.)

density

2.036 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

I[13C]([2H])([2H])[13C]([2H])([2H])[2H]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service