Skip to Content
Merck
All Photos(1)

Key Documents

691712

Sigma-Aldrich

7-Chloro-2-oxindole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6ClNO
CAS Number:
Molecular Weight:
167.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

221-225 °C

functional group

chloro

SMILES string

Clc1cccc2CC(=O)Nc12

InChI

1S/C8H6ClNO/c9-6-3-1-2-5-4-7(11)10-8(5)6/h1-3H,4H2,(H,10,11)

InChI key

FPDLUAACCNVSQA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Colgan et al.
Journal of pharmaceutical and biomedical analysis, 14(7), 825-833 (1996-05-01)
5-Chlorooxindole (5-CO) is a starting material for tenidap sodium, a pharmaceutical drug candidate produced by Pfizer. To insure potency and purity of the drug substance, it is necessary to demonstrate that commercial supplies of 5-CO are free from elevated levels

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service