Skip to Content
Merck
All Photos(1)

Key Documents

685887

Supelco

Sucrose solution

NMR reference standard, 2 mM in H2O:D2O (9:1) (99.9 atom % D), NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
Molecular Weight:
342.30
UNSPSC Code:
12142201
NACRES:
NA.24

grade

NMR reference standard

concentration

2 mM in H2O:D2O (9:1) (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

Looking for similar products? Visit Product Comparison Guide

General description

Sucrose is a disaccharide, naturally occurring carbohydrate found in plants and a major product during photosynthesis.[1][2]
Sucrose solution can be used as a water suppression standard in the determination of chemical composition of dissolved organic water in natural water bodies.[3]

Quantity

5 mm O.D. tube contains 0.700 mL.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct 1H NMR spectroscopy of dissolved organic matter in natural waters
Lam B and Simpson.JA
Analyst, 133, 263-269 (2008)
D. H. Lewis
Storage Carbohydrates in Vascular Plants: Distribution, Physiology and Metabolism, 53 -53 (1984)
Quantitative 1H NMR with external standards: use in preparation of calibration solutions for algal toxins and other natural products.
Burton IW, Quilliam MA, Walter JA.
Analytical Chemistry, 77(10), 3123-3131 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service