Skip to Content
Merck
All Photos(1)

Key Documents

685011

Sigma-Aldrich

(1,5-Cyclooctadiene)-η5-indenyl)iridium(I)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H7)Ir(C8H12)
CAS Number:
Molecular Weight:
414.54
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: iridium
reagent type: catalyst

mp

126-131 °C

SMILES string

[Ir].C1CC=CCCC=C1.[CH]2[CH][C]3C=CC=C[C]3[CH]2

InChI

1S/C9H7.C8H12.Ir/c1-2-5-9-7-3-6-8(9)4-1;1-2-4-6-8-7-5-3-1;/h1-7H;1-2,7-8H,3-6H2;/b;2-1-,8-7-;

InChI key

YLWHBOFZCLHZHQ-GHDUESPLSA-N

Application

Catalyst for C-H activation/borylation[1][2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Yang Cho et al.
Science (New York, N.Y.), 295(5553), 305-308 (2001-11-24)
Arylboron compounds have intriguing properties and are important building blocks for chemical synthesis. A family of Ir catalysts now enables the direct synthesis of arylboron compounds from aromatic hydrocarbons and boranes under "solventless" conditions. The Ir catalysts are highly selective
Robert E Maleczka et al.
Journal of the American Chemical Society, 125(26), 7792-7793 (2003-06-26)
An efficient one-pot C-H activation/borylation/oxidation protocol for the preparation of phenols is described. This method is particularly attractive for the generation of meta-substituted phenols bearing ortho-/para-directing groups, as such substrates are difficult to access by other phenol syntheses.

Articles

Arylboronic acids and esters, vital tools in chemical transformations, find extensive use, particularly in the Suzuki-Miyaura cross-coupling reaction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service