Skip to Content
Merck
All Photos(2)

Key Documents

683353

Sigma-Aldrich

Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)

greener alternative

Synonym(s):

(1,5-Cyclooctadiene)bis(2-methylallyl)ruthenium, Bis(2-methallyl)(1,5-cyclooctadiene)ruthenium, Ru(Me-allyl)2(COD), Ruthenium(II) bis(2-methylallyl) 1,5-cyclooctadiene complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ru(methylallyl)2(COD)
CAS Number:
Molecular Weight:
319.45
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

81.7-82.4 °C (lit.)

greener alternative category

storage temp.

−20°C

SMILES string

C1CC=CCCC=C1.CC(=C)C[Ru]CC(C)=C

InChI

1S/C8H12.2C4H7.Ru/c1-2-4-6-8-7-5-3-1;2*1-4(2)3;/h1-2,7-8H,3-6H2;2*1-2H2,3H3;/b2-1-,8-7-;;;

InChI key

GRYFGFSAXGLJLN-PHFPKPIQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

Legal Information

A product of Umicore

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuğçe Ayvalı et al.
Dalton transactions (Cambridge, England : 2003), 46(43), 15070-15079 (2017-10-27)
The choice of suitable organometallic precursors, [Re

Articles

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service