Skip to Content
Merck
All Photos(1)

Key Documents

682756

Sigma-Aldrich

1-(3-Bromo-1-propynyl)naphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9Br
CAS Number:
Molecular Weight:
245.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

43-47 °C

functional group

bromo

SMILES string

BrCC#Cc1cccc2ccccc12

InChI

1S/C13H9Br/c14-10-4-8-12-7-3-6-11-5-1-2-9-13(11)12/h1-3,5-7,9H,10H2

InChI key

LHDMQJTYLOVTPS-UHFFFAOYSA-N

Application

Reagent for the protection of alcohols in the form of a 1-naphthylpropargyl ether that can be cleaved with DDQ.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Crich et al.
Organic letters, 8(21), 4879-4882 (2006-10-06)
[reaction: see text] The (1-naphthyl)propargyl group is introduced as a sterically unobtrusive alcohol protecting group that is cleaved in a single step by exposure to dichlorodicyanoquinone in wet dichloromethane. In conjunction with the 4,6-O-benzylidene protecting group, and the use of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service