Skip to Content
Merck
All Photos(2)

Key Documents

675148

Sigma-Aldrich

5-Bromo-8-nitroisoquinoline

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5BrN2O2
CAS Number:
Molecular Weight:
253.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

90%

form

solid

mp

136-140 °C

functional group

bromo
nitro

SMILES string

[O-][N+](=O)c1ccc(Br)c2ccncc12

InChI

1S/C9H5BrN2O2/c10-8-1-2-9(12(13)14)7-5-11-4-3-6(7)8/h1-5H

InChI key

ULGOLOXWHJEZNZ-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service