Skip to Content
Merck
All Photos(1)

Documents

675121

Sigma-Aldrich

(3,3,3-Trichloropropyl)triphenylphosphonium chloride

95%

Synonym(s):

Triphenyl(3,3,3-Trichloropropyl)phosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCH2CH2P(C6H5)3Cl
CAS Number:
Molecular Weight:
444.16
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

138-145 °C

SMILES string

[Cl-].ClC(Cl)(Cl)CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H19Cl3P.ClH/c22-21(23,24)16-17-25(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20;/h1-15H,16-17H2;1H/q+1;/p-1

InChI key

CVLBDGPLEYFDOR-UHFFFAOYSA-M

Application

Reactant for:
  • Synthesis of DL-histrionicotoxin via a nine step synthesis involving cross-metathesis, oxime formation and Michael addition as the key steps
  • Wittig reactions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service