Skip to Content
Merck
All Photos(1)

Key Documents

672386

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium methyl carbonate solution

~50% in methanol: water (2:3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23NO3
CAS Number:
Molecular Weight:
217.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

concentration

~50% in methanol: water (2:3)

refractive index

n20/D 1.410

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

COC([O-])=O.CCCC[N+]1(C)CCCC1

InChI

1S/C9H20N.C2H4O3/c1-3-4-7-10(2)8-5-6-9-10;1-5-2(3)4/h3-9H2,1-2H3;1H3,(H,3,4)/q+1;/p-1

InChI key

JFACAAAJNPXLSG-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Eyes, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

121.1 °F

Flash Point(C)

49.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CBILS© offers a simple, elegant synthetic method for the production of ionic liquids, developed by the Austrian company proionic.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service