All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H10O
CAS Number:
Molecular Weight:
98.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
refractive index
n20/D 1.451
density
0.888 g/mL at 25 °C
SMILES string
CCC(O)C#CC
InChI
1S/C6H10O/c1-3-5-6(7)4-2/h6-7H,4H2,1-2H3
InChI key
KARLLBDFLHNKBO-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
127.9 °F
Flash Point(C)
53.3 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service