Skip to Content
Merck
All Photos(1)

Documents

664006

Sigma-Aldrich

Diethyl fluoro(phenylsulfonyl)methylphosphonate

97%

Synonym(s):

Diethyl [α-fluoro-α-(phenylsulfonyl)methyl]phosphonate, McCarthy′s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16FO5PS
CAS Number:
Molecular Weight:
310.28
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

mp

61-65 °C

SMILES string

CCOP(=O)(OCC)C(F)S(=O)(=O)c1ccccc1

InChI

1S/C11H16FO5PS/c1-3-16-18(13,17-4-2)11(12)19(14,15)10-8-6-5-7-9-10/h5-9,11H,3-4H2,1-2H3

InChI key

HUOWVYVTKZRXGM-UHFFFAOYSA-N

Application

Reactant for:
  • Radical-mediated thiodesulfonylation reactions
  • Preparation of Bacillus subtilis S-ribosylhomocysteinase inhibitors
  • Phenyl fluorovinyl sulfones via Julia olefination
  • Horner-Wadsworth-Emmons reaction
  • Negishi cross-coupling

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service