65946
tert-Butyl 3-butenoate
≥98.0% (GC)
Synonym(s):
tert-Butyl vinylacetate
About This Item
Recommended Products
Assay
≥98.0% (GC)
refractive index
n20/D 1.413
density
0.877 g/mL at 20 °C
SMILES string
CC(C)(C)OC(=O)CC=C
InChI
1S/C8H14O2/c1-5-6-7(9)10-8(2,3)4/h5H,1,6H2,2-4H3
InChI key
NGASWKRTXGWPNN-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
87.8 °F - closed cup
Flash Point(C)
31 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service