Skip to Content
Merck
All Photos(1)

Key Documents

657166

Sigma-Aldrich

2-Chloro-1,3,2-diazaphospholidine-2-sulfide, polymer-bound

100-200 mesh particle size, extent of labeling: 1-2 mmol/g loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 657166, please contact your local Merck office or dealer. Contact Technical Service

crosslinking

1 % cross-linked with divinylbenzene

extent of labeling

1-2 mmol/g loading

particle size

100-200 mesh

storage temp.

2-8°C

Application

Used for the conversion of primary amides to nitriles. Also converts secondary and tertiary amides to thioamides.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A polymer-supported thionating reagent.
Steven V. Ley, Andrew G. Leach and R. Ian Storer
Journal of the Chemical Society. Perkin Transactions 1, 358-361 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service