Skip to Content
Merck
All Photos(1)

Key Documents

639222

Sigma-Aldrich

Lithium 2-ethylhexoxide

10-13 wt. % in heptane

Synonym(s):

Lithium 2-ethyl-1-hexanolate, Lithium 2-ethyl-1-hexylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiOCH2CH(CH2CH3)(CH2)3CH3
CAS Number:
Molecular Weight:
136.16
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

concentration

10-13 wt. % in heptane
87-90% in heptanes

impurities

<2% 2-Ethyl-1-hexanol

SMILES string

[Li+].CCCCC(CC)C[O-]

InChI

1S/C8H17O.Li/c1-3-5-6-8(4-2)7-9;/h8H,3-7H2,1-2H3;/q-1;+1

InChI key

LWKRTMNGAUUVIN-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service