Skip to Content
Merck
All Photos(1)

Key Documents

638579

Sigma-Aldrich

Ethyl 2-bromo-(4-bromophenyl)acetate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
£52.50

£52.50

List Price£105.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
£52.50

About This Item

Empirical Formula (Hill Notation):
C10H10Br2O2
CAS Number:
Molecular Weight:
321.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

£52.50

List Price£105.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.5710 (lit.)

bp

140 °C/5 mmHg (lit.)

density

1.642 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

CCOC(=O)C(Br)c1ccc(Br)cc1

InChI

1S/C10H10Br2O2/c1-2-14-10(13)9(12)7-3-5-8(11)6-4-7/h3-6,9H,2H2,1H3

InChI key

PPRAWWBOUOBAON-UHFFFAOYSA-N

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daria Kupczyk et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-19)
Glucocorticoid metabolism at the tissue level is regulated by two isoenzymes 11β-hydroxysteroid dehydrogenase (11β-HSD), which mutually convert biologically active cortisol and inactive cortisone. Recent research is focused on the role of 11β-HSD1 and 11β-HSD2 as autocrine factors of tumor cell
Renata Studzińska et al.
Chemistry & biodiversity, 16(6), e1900065-e1900065 (2019-04-24)
11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) is an enzyme that affects the body's cortisol levels. The inhibition of its activity can be used in the treatment of Cushing's syndrome, metabolic syndrome and type 2 diabetes. In this study, we synthesized new

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service