Skip to Content
Merck
All Photos(1)

Key Documents

634166

Sigma-Aldrich

Iron(III) titanate

nanopowder

Synonym(s):

Iron(III) titanium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2TiO5
CAS Number:
Molecular Weight:
239.55
UNSPSC Code:
12352302
Pricing and availability is not currently available.

form

nanopowder

mean particle size

<60 nm (BET)

Features and Benefits

Nanopowder posseses interesting optical properties.[1]

Packaging

Packaged in glass bottles

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gracia, F. et al.
Surface and Coating Tech., 158-159, 552-552 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service