Skip to Content
Merck
All Photos(1)

Key Documents

633186

Sigma-Aldrich

Cetyl chloroformate

96%

Synonym(s):

Hexadecyl chloroformate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H33ClO2
CAS Number:
Molecular Weight:
304.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.4480 (lit.)

density

0.923 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCOC(Cl)=O

InChI

1S/C17H33ClO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20-17(18)19/h2-16H2,1H3

InChI key

HOQUWXSARQBQCW-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service