Skip to Content
Merck
All Photos(1)

Key Documents

61930

Sigma-Aldrich

L-tert-Leucinol hydrochloride

≥99.0% (CHN)

Synonym(s):

(S)-2-Amino-3,3-dimethyl-1-butanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO · HCl
Molecular Weight:
153.65
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (CHN)

optical activity

[α]20/D +35±1°, c = 1% in H2O

mp

196-198 °C

application(s)

peptide synthesis

SMILES string

Cl[H].CC(C)(C)[C@H](N)CO

InChI

1S/C6H15NO.ClH/c1-6(2,3)5(7)4-8;/h5,8H,4,7H2,1-3H3;1H/t5-;/m1./s1

InChI key

BFBMCXSYQJNKSS-NUBCRITNSA-N

Other Notes

Chiral auxiliary and building block which usually induces higher stereoselectivities than the corresponding derivatives of valinol; e.g. Claisen rearrangement of N-allyl ketene-N,O-acetals; preparation of an effective C2-symmetrical catalyst for hydrosilylation of ketones; alkylation of bicyclic N,O-acetals with keto acids; Ullmann coupling

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T.D. Nelson et al.
The Journal of Organic Chemistry, 59, 2655-2655 (1994)
M.J. Kurth et al.
Journal of the American Chemical Society, 107, 443-443 (1985)
H. Nishiyama et al.
Organometallics, 8, 846-846 (1989)
A.I. Meyers et al.
Tetrahedron Letters, 30, 1745-1745 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service