Skip to Content
Merck
All Photos(1)

Key Documents

618519

Sigma-Aldrich

Phosgene-12C solution

~1 M in benzene, 99.9 atom % 12C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
12COCl2
Molecular Weight:
98.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

description

13C-depleted

isotopic purity

99.9 atom % 12C

concentration

~1 M in benzene

mass shift

depleted

storage temp.

−20°C

SMILES string

Cl[12C](Cl)=O

InChI

1S/CCl2O/c2-1(3)4/i1+0

InChI key

YGYAWVDWMABLBF-IGMARMGPSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy J Herdendorf et al.
Biochemistry, 53(35), 5647-5660 (2014-08-20)
Spontaneous double-strand breaks (DSBs) are one of the most deleterious forms of DNA damage, and their improper repair can lead to cellular dysfunction. The Mre11 and Rad50 proteins, a nuclease and an ATPase, respectively, form a well-conserved complex that is
Junfeng Chen et al.
Inhalation toxicology, 27(5), 254-261 (2015-05-15)
Accidental phosgene exposure could result in acute lung injury (ALI), effective therapy is needed for the patients with phosgene-induced ALI. As a type of cells with therapeutic potential, mesenchymal stem cells (MSCs) have been showed its efficacy in multiple diseases.
Y Juillet et al.
Analytical and bioanalytical chemistry, 406(21), 5137-5145 (2014-05-13)
A new rapid, sensitive and reliable method was developed for the determination of phosgene in air samples using thermal desorption (TD) followed by gas chromatography-mass spectrometry (GC-MS). The method is based on a fast (10 min) active sampling of only
Hellmuth Lilienthal et al.
Toxicology, 327, 22-31 (2014-12-03)
Whereas the effects of dioxin-like polychlorinated biphenyls (DL-PCBs) are well described, less is known about non-dioxin-like PCBs (NDL-PCBs), including influences on the nervous system and related behavioral effects after developmental exposure. Following the examination of the highly purified NDL congeners

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service