Skip to Content
Merck
All Photos(3)

Key Documents

576689

Sigma-Aldrich

2-Iodo-4-nitroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(NO2)NH2
CAS Number:
Molecular Weight:
264.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

105-109 °C (lit.)

functional group

iodo
nitro

SMILES string

Nc1ccc(cc1I)[N+]([O-])=O

InChI

1S/C6H5IN2O2/c7-5-3-4(9(10)11)1-2-6(5)8/h1-3H,8H2

InChI key

LOLSEMNGXKAZBZ-UHFFFAOYSA-N

General description

2-Iodo-4-nitroaniline can be synthesized via reaction of 4-nitroaniline with iodine and silver acetate.[1]

Application

2-iodo-4-nitroaniline may be used to synthesize the following:
  • 2-iodo-4-nitrobenzonitrile via reaction with sodium nitrite to form a diazonium salt, which then reacts with a mixture of CuCN/KCN[2]
  • 2-iodo-p-phenylenediamine by reacting with tin(II)dihydrate in conc.HCl[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of optically active ring-A substituted tryptophans as IDO inhibitors"
Li X, et al.
Tetrahedron Letters, 45(46), 8569-8573 (2004)
"Preparation of 2-arylated-1, 4-phenylenediamines by palladium-catalyzed cross-coupling reactions"
Jensen EA, et al.
Journal of Organometallic Chemistry, 653(01), 122-128 (2002)
"Identification and optimization of a novel series of [2.2. 1]-oxabicyclo imide-based androgen receptor antagonists"
Salvati.EM, et al.
Bioorganic & Medicinal Chemistry Letters, 18(06), 1910-1915 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service