567957
3-Benzylphenyl isocyanate
97%
Synonym(s):
3′-Benzylphenyl isocyanate, 3′-Isocyanatodiphenylmethane
About This Item
Assay
97%
refractive index
n20/D 1.5880 (lit.)
density
1.106 g/mL at 25 °C (lit.)
SMILES string
O=C=Nc1cccc(Cc2ccccc2)c1
InChI
1S/C14H11NO/c16-11-15-14-8-4-7-13(10-14)9-12-5-2-1-3-6-12/h1-8,10H,9H2
InChI key
OUMFOCWUDYJGHI-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service