Skip to Content
Merck
All Photos(1)

Key Documents

566179

Sigma-Aldrich

Bis(pentafluorophenyl)borane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6F5)BH
CAS Number:
Molecular Weight:
345.93
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service