564044
4-Ethoxy-3-fluorophenylboronic acid
Synonym(s):
4-Ethoxy-3-fluorobenzeneboronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
solid
SMILES string
CCOc1ccc(cc1F)B(O)O
InChI
1S/C8H10BFO3/c1-2-13-8-4-3-6(9(11)12)5-7(8)10/h3-5,11-12H,2H2,1H3
InChI key
ZONJMULNQBNBGU-UHFFFAOYSA-N
Other Notes
Contains varying amounts of anhydride
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service