Skip to Content
Merck
All Photos(1)

Key Documents

552798

Sigma-Aldrich

Gallium(III) chloride solution

0.5 M in pentane

Synonym(s):

Gallium chloride, Gallium trichloride, Trichlorogallium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaCl3
CAS Number:
Molecular Weight:
176.08
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

reagent type: catalyst
core: gallium

concentration

0.5 M in pentane

bp

35 °C

density

0.688 g/mL at 25 °C

SMILES string

Cl[Ga](Cl)Cl

InChI

1S/3ClH.Ga/h3*1H;/q;;;+3/p-3

InChI key

UPWPDUACHOATKO-UHFFFAOYSA-K

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-26.0 °F - closed cup

Flash Point(C)

< -32.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kee-Lung Chang et al.
Toxicology and applied pharmacology, 193(2), 209-217 (2003-12-03)
Gallium is commonly used in the semiconductor industry and medical field. Biologically, gallium is able to interrupt iron metabolism. Exposure to gallium has been shown to affect the human immune system. The purpose of this study was to investigate the
Cristina Nanni et al.
Clinical physiology and functional imaging, 29(3), 187-192 (2009-03-27)
[(18)F]-FDG is a widely used tracer for the non-invasive evaluation of hypermetabolic processes like cancer and inflammation. However, [(18)F]-FDG is considered inaccurate for the diagnosis of urinary tract and genital infections because of its urinary excretion. Since the 1970s, Gallium
E M Perchellet et al.
Anti-cancer drugs, 10(5), 477-488 (1999-09-07)
Gallium chloride (GaCl3), an antitumor agent with antagonistic action on iron, magnesium and calcium, was tested for its ability to alter the polymerization of purified tubulin (2.2 mg/ml) in a cell-free system in vitro. GaCl3 (250 microM) does not mimic
João Batista Borges et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 52(2), 206-209 (2011-01-15)
Ventilation distribution can be assessed by SPECT with Technegas. This study was undertaken in piglets with different degrees of ventilation inhomogeneity to compare PET using (68)Ga-labeled pseudogas or "Gallgas" with Technegas. Twelve piglets were studied in 3 groups: control, lobar
Acute and chronic effects of gallium chloride (GaCl3) on tilapia (Oreochromis mossambicus) larvae.
H C Lin et al.
Bulletin of environmental contamination and toxicology, 60(6), 931-935 (1998-06-12)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service