Skip to Content
Merck
All Photos(1)

Key Documents

550930

Sigma-Aldrich

1-(2-Chlorophenyl)-3-buten-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH(OH)CH2CH=CH2
CAS Number:
Molecular Weight:
182.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

mp

31-35 °C (lit.)

SMILES string

OC(CC=C)c1ccccc1Cl

InChI

1S/C10H11ClO/c1-2-5-10(12)8-6-3-4-7-9(8)11/h2-4,6-7,10,12H,1,5H2

InChI key

BFIIUQFVPDIOLQ-UHFFFAOYSA-N

General description

1-(2-Chlorophenyl)-3-buten-1-ol can be prepared by the allylation of o-benzaldehyde with allyl bromide, mediated by tin/iodine system in water.[1]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sn/I2 Mediated allylation of carbonyl compounds with allyl (crotyl) halide in water.
Zhou Y, et al.
ARKIVOC (Gainesville, FL, United States), 11, 142-153 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service