Skip to Content
Merck
All Photos(1)

Key Documents

550434

Sigma-Aldrich

Diethyl selenide

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Se(C2H5)2
CAS Number:
Molecular Weight:
137.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.999% trace metals basis

form

liquid

bp

108 °C (lit.)

density

1.232 g/mL at 25 °C (lit.)

SMILES string

CC[Se]CC

InChI

1S/C4H10Se/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

ALCDAWARCQFJBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 34 Selenium

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

72.0 °F - closed cup

Flash Point(C)

22.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gustavo Moreno-Martin et al.
Talanta, 224, 121907-121907 (2021-01-01)
Quantification of volatile organoselenium species released by Escherichia coli (E. coli) and Staphylococcus aureus (S. aureus), after their growth in the presence of 1 and 2 mg Se·L-1 as both selenite and chitosan-modified selenium nanoparticles (Ch-SeNPs), was achieved by the application
Fernando Gabriel Martínez et al.
Frontiers in bioengineering and biotechnology, 8, 506-506 (2020-07-01)
Selenium (Se) is an essential micronutrient for the majority of living organisms, and it has been identified as selenocysteine in the active site of several selenoproteins such as glutathione peroxidase, thioredoxin reductase, and deiodinases. Se deficiency in humans is associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service