Skip to Content
Merck
All Photos(1)

Key Documents

549762

Sigma-Aldrich

Ethyl 6,8-dichlorooctanoate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OCO(CH2)4CH(Cl)(CH2)2Cl
CAS Number:
Molecular Weight:
241.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.462 (lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCCC(Cl)CCCl

InChI

1S/C10H18Cl2O2/c1-2-14-10(13)6-4-3-5-9(12)7-8-11/h9H,2-8H2,1H3

InChI key

RFYDWSNYTVVKBR-UHFFFAOYSA-N

Application

Ethyl 6,8-dichlorooctanoate may be used as a starting material in the synthesis of 6-selenolipoic acid and α-lipoic acid derivatives, which show potent anticancer activity.
It may also be used as a starting material to synthesize:
  • 6,8-Dibenzylmercaptooctanoic acid, an intermediate for the preparation of DL-α-lipoic acid.
  • Ethyl 5,7-dichloroheptanoate, an intermediate for the preparation of DL-1,2-dithiolane-3-butyric acid.
  • 7-Bromo-1,3-dichloroheptane, an intermediate for the preparation of DL-12-dithiolane-3-butanesulfonamide.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of some monoselenolipoic acid derivatives and their biological evaluation as anticancer agents.
Xu F, et al.
J. Chem. Res. (M), 37(5), 311-314 (2013)
Syntheses of DL-a-lipoic acid.
Reed LJ and Niu CI.
Journal of the American Chemical Society, 77(2), 416-419 (1955)
Synthesis of DL-1, 2-Dithiolane-3-caproic Acid and DL-1, 2-Dithiolane-3-butyric Acid, Homologs of a-Lipoic Acid.
Thomas RC and Reed LJ.
Journal of the American Chemical Society, 78(23), 6151-6153 (1956)
Synthesis of DL-1, 2-Dithiolane-3-butanesulfonamide, an Analog of a-Lipoic Acid.
Thomas RC and Reed LJ.
Journal of the American Chemical Society, 78(23), 6150-6151 (1956)
Shi-Jie Zhang et al.
Bioorganic & medicinal chemistry letters, 20(10), 3078-3083 (2010-04-21)
alpha-Lipoic acid derivatives were synthesized and evaluated for their in vitro anticancer activities against NCI-460, HO-8910, KB, BEL-7402, and PC-3 cell lines. The results, for most compounds exhibited dose-dependent inhibitory property and several compounds had good inhibitions at the dose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service