Skip to Content
Merck
All Photos(1)

Key Documents

545287

Sigma-Aldrich

Ethyl trans-2-hydroxycinnamate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OH)CH=CHCO2C2H5
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

84-87 °C (lit.)

SMILES string

[H]\C(=C(\[H])c1ccccc1O)C(=O)OCC

InChI

1S/C11H12O3/c1-2-14-11(13)8-7-9-5-3-4-6-10(9)12/h3-8,12H,2H2,1H3/b8-7+

InChI key

SRBYHEKXHQYMRN-BQYQJAHWSA-N

General description

Ethyl trans-2-hydroxycinnamate is an o-hydroxy cinnmate ester derivative.

Application

Ethyl trans-2-hydroxycinnamate [(E)-ethyl 3-(2-hydroxyphenyl) acrylate] may be used to prepare 4-phenyl-2H-chromen-2-one, a 4-aryl coumarin derivative.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 4-arylcoumarins via palladium-catalyzed arylation/cyclization of ortho-hydroxylcinnamates with diaryliodonium salts.
Yang Y, et al.
Tetrahedron Letters, 56(24), 3809-3812 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service