Skip to Content
Merck
All Photos(1)

Key Documents

539783

Sigma-Aldrich

5-(4-Chlorophenyl)-1,3-cyclohexanedione

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4C6H7O2
CAS Number:
Molecular Weight:
222.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

185-189 °C (lit.)

functional group

chloro
ketone

SMILES string

Clc1ccc(cc1)C2CC(=O)CC(=O)C2

InChI

1S/C12H11ClO2/c13-10-3-1-8(2-4-10)9-5-11(14)7-12(15)6-9/h1-4,9H,5-7H2

InChI key

MAFYKXZPGMHTIA-UHFFFAOYSA-N

Application

5-(4-Chlorophenyl)-1,3-cyclohexanedione may be used in the synthesis of 3-(5-(4-chlorophenyl)-3-oxocyclohex-1-enylamino)thiophene-2-carbonitrile.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 9-amino-5, 6, 7, 8-tetrahydrothieno [3, 2-b] quinoline and 9-amino-5, 6, 7, 8-tetrahydrothieno [3, 2-b] quinolin-8-ol derivatives.
Song YH and Jo BS.
Journal of Heterocyclic Chemistry, 46(6), 1132-1136 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service