Skip to Content
Merck
All Photos(3)

Key Documents

532045

Sigma-Aldrich

Methyl 4,4-dimethoxybutyrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CH(CH2)2CO2CH3
CAS Number:
Molecular Weight:
162.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.417 (lit.)

bp

40-50 °C/0.2 mmHg (lit.)

density

1.05 g/mL at 25 °C (lit.)

functional group

acetal
ester
ether

SMILES string

COC(CCC(=O)OC)OC

InChI

1S/C7H14O4/c1-9-6(8)4-5-7(10-2)11-3/h7H,4-5H2,1-3H3

InChI key

MMRXJPOAPCEDLP-UHFFFAOYSA-N

General description

Methyl 4,4-dimethoxybutyrate (methyl 4,4-dimethoxybutanoate) can be prepared by the electrochemical ring-opening of furan-2-carboxylic acid in the presence of methanol and using graphite electrode.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Warriner SL.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 29, 371-371 (2014)
Cordell GA.
The Alkaloids, 52, 141-141 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service