Skip to Content
Merck
All Photos(1)

Documents

531650

Sigma-Aldrich

5-Nitrofurfurylnitrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O6
CAS Number:
Molecular Weight:
188.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

36-39 °C (lit.)

SMILES string

[O-][N+](=O)OCc1ccc(o1)[N+]([O-])=O

InChI

1S/C5H4N2O6/c8-6(9)5-2-1-4(13-5)3-12-7(10)11/h1-2H,3H2

InChI key

IZUDEUAXDJRXOA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Masek et al.
Mutation research, 281(1), 63-66 (1992-01-01)
Inducible stable DNA replication (iSDR) provoked by a damaging treatment with MMS, MNU, MNNG, NFAA, NFN, 4NQO, NAL or MMC, was followed in both repair-competent E. coli PQ35 and its uvrA derivative E. coli PQ37. In contrast to SOS-inducible mutagenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service