Skip to Content
Merck
All Photos(1)

Key Documents

525731

Sigma-Aldrich

3,4,5-Triethoxybenzyl alcohol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3C6H2CH2OH
CAS Number:
Molecular Weight:
240.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

96%

mp

36-39 °C (lit.)

SMILES string

CCOc1cc(CO)cc(OCC)c1OCC

InChI

1S/C13H20O4/c1-4-15-11-7-10(9-14)8-12(16-5-2)13(11)17-6-3/h7-8,14H,4-6,9H2,1-3H3

InChI key

BDLRHPZTSRLNOW-UHFFFAOYSA-N

General description

3,4,5-Triethoxybenzyl alcohol is an organic building block useful in organic synthesis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service