Skip to Content
Merck
All Photos(1)

Key Documents

525561

Sigma-Aldrich

6-Bromo-2-phenyl-(4H)-4-benzopyranone

97%

Synonym(s):

6-Bromo-2-phenyl-4H-1-benzopyran-4-one, 6-Bromoflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9BrO2
CAS Number:
Molecular Weight:
301.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

193-196 °C (lit.)

SMILES string

Brc1ccc2OC(=CC(=O)c2c1)c3ccccc3

InChI

1S/C15H9BrO2/c16-11-6-7-14-12(8-11)13(17)9-15(18-14)10-4-2-1-3-5-10/h1-9H

InChI key

LPOZDKMXSPAATL-UHFFFAOYSA-N

Gene Information

General description

6-Bromo-2-phenyl-(4H)-4-benzopyranone is also referred to as 6-bromoflavone. It is a mononuclear halogenated flavone.[1] 6-Bromoflavone can be prepared from flavanone, via bromination.[2] It can also be synthesized from 2′-hydroxychalcone dibromide.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of 2'-hydroxychalcone dibromides with pyridine.
Reddy NJ, et al.
The Journal of Organic Chemistry, 46(3), 638-641 (1981)
6-Bromoflavone, a high affinity ligand for the central benzodiazepine receptors is a member of a family of active flavonoids.
Marder M, et al.
Biochemical and Biophysical Research Communications, 223(2), 384-389 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service