Skip to Content
Merck
All Photos(1)

Key Documents

525049

Sigma-Aldrich

4-Methoxy-2-methylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 525049, please contact your local Merck office or dealer. Contact Technical Service

Assay

97%

mp

177-181 °C (lit.)

functional group

carboxylic acid

SMILES string

COc1ccc(C(O)=O)c(C)c1

InChI

1S/C9H10O3/c1-6-5-7(12-2)3-4-8(6)9(10)11/h3-5H,1-2H3,(H,10,11)

InChI key

MSVRGYOYISBGTH-UHFFFAOYSA-N

General description

4-Methoxy-2-methylbenzoic acid (2-methylanisic acid)[1] is a trisubstituted aromatic compound. It is a positional isomeric form of 2-methoxy-4-methylbenzoic acid.[2] 4-Methoxy-2-methylbenzoic acid readily forms complexes with Mn(II), Co(II), Ni(II), Cu(II), Zn(II) and Cd(II).[2]

Application

4-Methoxy-2-methylbenzoic acid may be used in the synthesis of 4-methoxy-2-methylbenzoates of Y(III) and lanthanides (III) (La to Lu, excluding Pm).[1] It may also be used to synthesize 4-methoxy-2-methylbenzoic acid hydrazide[3] and 5-bromo-4-methoxy-2-methylbenzoic acid.[4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal, spectral and magnetic investigations of Mn (II), Co (II), Ni (II), Cu (II), Zn (II) and cd (ii) complexes with 4-methoxy-2-methylbenzoic acid.
Meganathan C, et al.
Polish Journal of Chemistry, 76(12), 1689-1692 (2002)
(1 S)-1, 5-Anhydro-1-[5-(4-ethoxybenzyl)-2-methoxy-4-methylphenyl]-1-thio-d-glucitol (TS-071) is a potent, selective sodium-dependent glucose cotransporter 2 (SGLT2) inhibitor for type 2 diabetes treatment.
Kakinuma H, et al.
Journal of Medicinal Chemistry, 58(3), 3247- 3261 (2010)
Spectral and thermal investigation of rare earth element 4-methoxy-2-methylbenzoates.
Brzyska W and Ozga W.
Journal of Thermal Analysis and Calorimetry, 70(2), 467-474 (2002)
A Omodei-Salé et al.
Journal of medicinal chemistry, 26(8), 1187-1192 (1983-08-01)
A series of 3,5-diaryl-s-triazoles were synthesized and evaluated as postimplantation contragestational agents. The introduction of various substituents (e.g., an o-alkyl chain on one phenyl and a m-alkoxy group on the other) was found to increase the potency. Several compounds with
Peng Hu et al.
Organic letters, 11(11), 2341-2344 (2009-05-13)
A versatile palladium catalyst system was developed to effect the decarboxylative Heck coupling of a variety of arenecarboxylic acids with a wide range of olefins. The key to obtaining the efficient catalyst system is the use of 1-adamatanecarboxylic acid as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service