Skip to Content
Merck
All Photos(1)

Key Documents

524751

Sigma-Aldrich

2-Methoxy-4-nitrobenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)CN
CAS Number:
Molecular Weight:
178.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

177-180 °C (lit.)

functional group

nitrile
nitro

SMILES string

COc1cc(ccc1C#N)[N+]([O-])=O

InChI

1S/C8H6N2O3/c1-13-8-4-7(10(11)12)3-2-6(8)5-9/h2-4H,1H3

InChI key

MLIKCKXLGYEGAO-UHFFFAOYSA-N

General description

2-Methoxy-4-nitrobenzonitrile is an organic building block.

Application

2-Methoxy-4-nitrobenzonitrile may be used in the preparation of 4-amino-2-methoxybenzonitrile.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akiyoshi Numadate et al.
Chemical & pharmaceutical bulletin, 62(10), 979-988 (2014-10-03)
An established inhibitor of dynamin-related protein 1 (Drp1), 3-(2,4-dichloro-5-methoxyphenyl)-2-thioxoquinazoline-4-one (mdivi-1), was recently reported also to show potent puromycin-sensitive aminopeptidase (PSA)-inhibitory activity. Herein, we report structural development of mdivi-1 derivatives and structure-activity relationship (SAR) analysis of the synthesized compounds, as well

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service