Skip to Content
Merck
All Photos(1)

Key Documents

514217

Sigma-Aldrich

4-(Octyloxy)aniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OC6H4NH2
CAS Number:
Molecular Weight:
221.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Product 514217 is not currently sold in your country. Contact Technical Service

Assay

98%

bp

336 °C (lit.)

mp

35-39 °C (lit.)

SMILES string

CCCCCCCCOc1ccc(N)cc1

InChI

1S/C14H23NO/c1-2-3-4-5-6-7-12-16-14-10-8-13(15)9-11-14/h8-11H,2-7,12,15H2,1H3

InChI key

ACYGZCHBIGKPGR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 612-612 (2014)
Woosung Kwon et al.
Scientific reports, 5, 12604-12604 (2015-07-29)
Carbon nanodots (C-dots) are a kind of fluorescent carbon nanomaterials, composed of polyaromatic carbon domains surrounded by amorphous carbon frames, and have attracted a great deal of attention because of their interesting properties. There are still, however, challenges ahead such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service