Skip to Content
Merck
All Photos(1)

Key Documents

510521

Sigma-Aldrich

1-Benzyloxy-3-methyl-2-nitrobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H3(CH3)NO2
CAS Number:
Molecular Weight:
243.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.5765 (lit.)

bp

150 °C/1.5 mmHg (lit.)

density

1.738 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(OCc2ccccc2)c1[N+]([O-])=O

InChI

1S/C14H13NO3/c1-11-6-5-9-13(14(11)15(16)17)18-10-12-7-3-2-4-8-12/h2-9H,10H2,1H3

InChI key

ZUMJNYJMXBJJIC-UHFFFAOYSA-N

General description

1-Benzyloxy-3-methyl-2-nitrobenzene is a substituted nitrobenzene.

Application

1-Benzyloxy-3-methyl-2-nitrobenzene may be used in the multi-step preparation of 7-hydroxy-oxindole-3-acetic acid.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Midoh et al.
Bioscience, biotechnology, and biochemistry, 74(9), 1794-1801 (2010-09-14)
The components contributing to the antioxidative activity of supersweet corn powder (SSCP), which is commonly used in corn soup and snacks in Japan, were clarified and the effects investigated. 7-(O-β-Glucosyloxy)oxindole-3-acetic acid (GOA) was found to be the component most strongly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service