Skip to Content
Merck
All Photos(4)

Key Documents

510211

Sigma-Aldrich

Nafion perfluorinated resin solution

greener alternative

5 wt. % in mixture of lower aliphatic alcohols and water, contains 45% water

Synonym(s):

D-521

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
eCl@ss:
32110203

eq. wt.

1,100

contains

45% water

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

5 wt. % in mixture of lower aliphatic alcohols and water

refractive index

n20/D 1.368

bp

84 °C

density

0.921 g/mL at 25 °C

greener alternative category

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Nafion perfluorinated resin solution is widely used as an electrolytic material. It can be used in the formation of a composite membrane with metal oxides. The membrane further facilitates the formation of chemically stable proton exchange membrane (PEM) fuel cells. It may be coated on mesoporous metallic films to improve the detection and selectivity of biomarkers. It may also form a composite electrode material with graphene for glucose biosensors.[1][2][3][4][5]
Support material for an integrated system which used solar energy to drive chemical reactions. Acts as a superacid catalyst for organic synthesis. When formed as metal composites, can be used as artificial muscle actuators, solid electrolytes for the analysis of electroactive species in organic media, or as electrodes in organic electrosynthesis.

Packaging

Packaged in glass bottles

Physical form

Hydrogen ion form

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuya Takeuchi et al.
Journal of bioscience and bioengineering, 123(3), 358-363 (2016-11-08)
Cellulomonas fimi, Cellulomonas biazotea, and Cellulomonas flavigena are cellulose-degrading microorganisms chosen to compare the degradation of cellulose. C. fimi degraded 2.5 g/L of cellulose within 4 days, which was the highest quantity among the three microorganisms. The electric current generation by the
Nichanan Phansroy et al.
Journal of bioscience and bioengineering, 126(4), 514-521 (2018-05-17)
A single chamber type microbial fuel cell (MFC) with 100 mL of chamber volume and 50 cm2 of air-cathode was developed in this study wherein a developed iron-plated carbon-felt anode and Shewanella oneidensis MR-1 were used. The performance of the iron-plated carbon-felt
An enzymatic glucose biosensor based on a glassy carbon electrode modified with manganese dioxide nanowires
Zhang L, et al.
Microchimica Acta, 54-56 (2013)
Glucose biosensor of ruthenium-doped TiO2 sensing electrode by co-sputtering system
Chou JC, et al.
Microelectronics reliability (2010)
Waller, F.J.; Van Scoyoc, R.W.
Chemtech, 17, 438-438 (1987)

Articles

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Related Content

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service