Skip to Content
Merck
All Photos(1)

Key Documents

50705

Sigma-Aldrich

Ethyl glyoxalate solution

technical, ~50% in toluene

Synonym(s):

Ethyl oxoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOCOOCH2CH3
CAS Number:
Molecular Weight:
102.09
Beilstein:
1209486
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical

concentration

~50% in toluene

density

1.03 g/mL at 20 °C

SMILES string

CCOC(=O)C=O

Caution

exists partly in the polymerized form; depolymerizes on heating

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service