Skip to Content
Merck
All Photos(1)

Key Documents

494089

Sigma-Aldrich

2-Fluoro-5-formylbenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CHO)CN
CAS Number:
Molecular Weight:
149.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

80-84 °C (lit.)

functional group

aldehyde
fluoro
nitrile

SMILES string

[H]C(=O)c1ccc(F)c(c1)C#N

InChI

1S/C8H4FNO/c9-8-2-1-6(5-11)3-7(8)4-10/h1-3,5H

InChI key

MOFRJTLODZILCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Fluoro-5-formylbenzonitrile is a benzonitrile derivative.

Application

2-Fluoro-5-formylbenzonitrile may be used in the synthesis of 3-cyano-4-fluorobenzyl bromide.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved synthesis of 3-cyano-4-fluorobenzyl bromide.
Cain GA.
ChemInform, 31(26) (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service