Skip to Content
Merck
All Photos(1)

Documents

492566

Sigma-Aldrich

Tris(cyclopentadienyl)gadolinium(III)

99.9% trace metals basis

Synonym(s):

GdCp3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd(C5H5)3
CAS Number:
Molecular Weight:
352.53
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

solid

reaction suitability

core: gadolinium
reagent type: catalyst

mp

295 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Gd].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.[CH]3[CH][CH][CH][CH]3

InChI

1S/3C5H5.Gd/c3*1-2-4-5-3-1;/h3*1-5H;

InChI key

ZCFWIGYKHNDYOA-UHFFFAOYSA-N

General description

Atomic number of base material: 64 Gadolinium

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2 - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service