Skip to Content
Merck
All Photos(1)

Key Documents

479128

Sigma-Aldrich

2-Aminoindan

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 479128, please contact your local Merck office or dealer. Contact Technical Service

Assay

97%

refractive index

n20/D 1.561 (lit.)

density

1.024 g/mL at 25 °C (lit.)

SMILES string

NC1Cc2ccccc2C1

InChI

1S/C9H11N/c10-9-5-7-3-1-2-4-8(7)6-9/h1-4,9H,5-6,10H2

InChI key

LMHHFZAXSANGGM-UHFFFAOYSA-N

General description

2-Aminoindan (2-Aminoindane) is an analog of amphetamine. It shows a potential bronchodilator and analgesic effect.[1] The impact of the intramolecular N-H···Π hydrogen bonding on the conformations of 2-Al has been analyzed.[2]

Application

2-Aminoindan may be used to prepare trans-(1S,2S)-2-amino-1-indanol via hydroxylation using dopamine β-hydroxylase (DBH) enzyme.[3] It may be used to synthesize 2-amino-4-chloro-6-(2-aminoindanyl)-1,3,5-triazine.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Hajduk et al.
Journal of medicinal chemistry, 42(19), 3852-3859 (1999-10-03)
The Erm family of methyltransferases confers resistance to the macrolide-lincosamide-streptogramin type B (MLS) antibiotics through the methylation of 23S ribosomal RNA. Upon the methylation of RNA, the MLS antibiotics lose their ability to bind to the ribosome and exhibit their
Hiroshi Iga et al.
The journal of physical chemistry. A, 111(27), 5981-5987 (2007-06-19)
Laser-induced fluorescence (LIF), dispersed fluorescence (DF), mass-resolved one-color resonance enhanced two-photon ionization (RE2PI) and UV-UV hole-burning spectra of 2-aminoindan (2-AI) were measured in a supersonic jet. The hole-burning spectra demonstrated that the congested vibronic structures observed in the LIF excitation
Adam L Halberstadt et al.
Psychopharmacology, 236(3), 989-999 (2019-03-25)
Over the last decade, many new psychostimulant analogues have appeared on the recreational drug market and most are derivatives of amphetamine or cathinone. Another class of designer drugs is derived from the 2-aminoindan structural template. Several members of this class
Enzymatic Hydroxylation by Dopamine β-Hydroxylase.
Mitrochkine AA, et al.
European Journal of Organic Chemistry, 1998(6), 1171-1176 (1998)
H Wikström et al.
Journal of medicinal chemistry, 30(7), 1115-1120 (1987-07-01)
Conformational analyses on differently substituted 2-aminoindans of significant pharmacological interest were carried out by the molecular mechanics method (MM2). An X-ray structure of (R)-4-methoxy-2-aminoindan has shown the ammonium nitrogen [-)-D-tartaric acid salt) in an axial position. From comparison with other

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service